missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ANXA2 (Human) Recombinant Protein
Human ANXA2 full-length ORF ( AAH66955, 19 a.a. - 357 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000302-P02.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSTVHEILCKLSLEGDHSTPPSAYGSVKAYTNFDAERDALNIETAIKTKGVDEVTIVNILTNRSNAQRQDIAFAYQRRTKKELASALKSALSGHLETVILGLLKTPAQYDASELKASMKGLGTDEDSLIEIICSRTNQELQEINRVYKEMYKTDLEKDIISDTSGDFRKLMVALAKGRRAEDGSVIDYELIDQDARDLYDAGVKRKGTDVPKWISVMTERSVPHLQKVFDRYKSYSPYDMLESIRKEVKGDLENAFLNLVQCIQNKPLYFADRLYDSMKGKGTRDKVLIRIMVSRSEVDMLKIRSEFKRKYGKSLYYYIQQDTKGDYQKALLYLCGGDD
Specifications
AAH66955 | |
annexin A2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANXA2 | |
GST |
302 | |
Wheat germ expression system | |
25 μg | |
ANX2, ANX2L4, CAL1H, LIP2, LPC2, LPC2D, P36, PAP-IV | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |