missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Abnova™ ANXA2 (Human) Recombinant Protein
Klik for at se tilgængelige muligheder
Mængde:
10 μg
25 μg
Pakningsstørrelse:
25µg
Beskrivelse
- Sequence: MSTVHEILCKLSLEGDHSTPPSAYGSVKAYTNFDAERDALNIETAIKTKGVDEVTIVNILTNRSNAQRQDIAFAYQRRTKKELASALKSALSGHLETVILGLLKTPAQYDASELKASMKGLGTDEDSLIEIICSRTNQELQEINRVYKEMYKTDLEKDIISDTSGDFRKLMVALAKGRRAEDGSVIDYELIDQDARDLYDAGVKRKGTDVPKWISVMTERSVPHLQKVFDRYKSYSPYDMLESIRKEVKGDLENAFLNLVQCIQNKPLYFADRLYDSMKGKGTRDKVLIRIMVSRSEVDMLKIRSEFKRKYGKSLYYYIQQDTKGDYQKALLYLCGGDD
Tekniske data
Tekniske data
| Adgangsnummer | AAH66955 |
| Gen-id (Entrez) | 302 |
| Navn | annexin A2 |
| Fremstillingsmetode | Wheat germ expression system |
| Kvalitetskontrol test | 125% SDS-PAGE Stained with Coomassie Blue |
| Mængde | 10 μg |
| Opbevaringskrav | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | ANX2, ANX2L4, CAL1H, LIP2, LPC2, LPC2D, P36, PAP-IV |
| Gen symbol | ANXA2 |
| Arter | Wheat Germ (in vitro) |
| Vis mere |
Korrektion af produktindhold
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction