Få mere at vide
Anle138b, MedChemExpress
Artikelnummer. 16492786
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16492786 25 mg
16476337 5 mg
16450007 50 mg
16420946 200 mg
16414706 100 mg
16413618 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 16492786 Leverandør Medchem Express Leverandørnr. HY101855
MedChemExpress Anle138b, an oligomeric aggregation inhibitor, blocks the formation of pathological aggregates of prion protein (PrPSc) and of α-synuclein (α-syn).
Anle138b strongly inhibits oligomer accumulation, neuronal degeneration, and disease progression in vivo. Anle138b has low toxicity and an excellent oral bioavailability and blood-brain-barrier penetration. Anle138b blocks Aβ channels and rescues disease phenotypes in a mouse model for amyloid pathology.
Tekniske data
Kemisk navn eller materiale Anle138b
CAS 882697-00-9
Farve Light Nude
Renhedsgrad noter Research
Molekylær formel C16H11BrN2O2
Mængde 25 mg
Opløselighedsinformation DMSO : ≥ 50 mg/mL (145.70 mM)
SMIL BrC1=CC(C2=CC(C3=CC=C(OCO4)C4=C3)=NN2)=CC=C1
Molekylvægt (g/mol) 343.17
Formel vægt 343.17
Procent renhed 99.18%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neurodegeneration