Få mere at vide
Amylocaine Hydrochloride, TRC
Artikelnummer. 30132923
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132923 25 mg
30012229 5 mg
30006991 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132923 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63670025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 532-59-2
Molekylær formel C14 H21 N O2 . Cl H
Molekylvægt (g/mol) 271.78
Synonym 2-Butanol, 1-(dimethylamino)-2-methyl-, 2-benzoate, hydrochloride (1:1), Amylocaine Hydrochloride, 2-Butanol, 1-(dimethylamino)-2-methyl-, benzoate (ester), hydrochloride (8CI,9CI), 2-Dimethylamino-1-ethyl-1-methylethyl benzoate hydrochloride, Amyleine hydrochloride, Amylocaine hydrochloride, Stovaine hydrochloride
IUPAC navn [1-(dimethylamino)-2-methylbutan-2-yl] benzoate;hydrochloride
SMIL Cl.CCC(C)(CN(C)C)OC(=O)c1ccccc1
Tekniske data
Kemisk navn eller materiale Amylocaine Hydrochloride
Analyt- eller komponentnavne Amylocaine Hydrochloride
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C14H21NO2.ClH/c1-5-14(2,11-15(3)4)17-13(16)12-9-7-6-8-10-12;/h6-10H,5,11H2,1-4H3;1H
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 25 mg
Formel vægt 271.1339
Procent renhed >95%