CAS RN 532-59-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Amylocaine Hydrochloride, TRC
CAS: 532-59-2 Molekylær formel: C14 H21 N O2 . Cl H Molekylvægt (g/mol): 271.78 Synonym: 2-Butanol, 1-(dimethylamino)-2-methyl-, 2-benzoate, hydrochloride (1:1),Amylocaine Hydrochloride,2-Butanol, 1-(dimethylamino)-2-methyl-, benzoate (ester), hydrochloride (8CI,9CI),2-Dimethylamino-1-ethyl-1-methylethyl benzoate hydrochloride,Amyleine hydrochloride,Amylocaine hydrochloride,Stovaine hydrochloride IUPAC navn: [1-(dimethylamino)-2-methylbutan-2-yl] benzoate;hydrochloride SMIL: Cl.CCC(C)(CN(C)C)OC(=O)c1ccccc1