missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMG 517

Description
Potent TRPV1 antagonist (IC50 = 0.9 nM). Inhibits capsaican-induced flinching and reverses analgesia in an inflammatory pain model in rats. Orally bioavailable.
Specifications
Specifications
| Kemisk navn eller materiale | N-[4-[[6-[4-(Trifluoromethyl)phenyl]-4-pyrimidinyl]oxy]-2-benzothiazolyl]acetamide |
| Mængde | 10 mg |
| Opløselighed | Soluble to 100mM in DMSO |
| Formel vægt | 430.41 |
| Renhed | >98% |
| Molekylvægt (g/mol) | 430.405 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?