Få mere at vide
Ambroxol hydrochloride, 98%, Thermo Scientific™
369.65 DKK - 1269.65 DKK
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15455899
|
Thermo Scientific Alfa Aesar
J63712.03 |
409.71 DKK 369.65 DKK / 1g Spare 40.06 DKK 9% Slukket |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | ||||||
|
15465899
|
Thermo Scientific Alfa Aesar
J63712.06 |
5g |
1410.47 DKK 1269.65 DKK / 5g Spare 140.82 DKK 9% Slukket |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsAmbroxol hydrochloride is a medication indicated to alleviate chest congestion associated with conditions that include bronchitis, pneumonia, bronchospasm asthma, cough, and allergy. Preclinically, ambroxol, the active ingredient of Mucosolvan, has been shown to increase respiratory tract secretion. It enhances pulmonary surfactant production and stimulates ciliary activity.
Solubility
Slightly soluble in water (∼5 mg/ml) or ethanol (∼5 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.