Learn More
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers, with Certificate
Amber glass to protect light-sensitive contents. Delivered with individual USP conformity certificate.
518.00 DKK - 692.00 DKK
Specifications
Closure Material | Polypropylene |
---|---|
Material | Borosilicate Glass (3.3) |
No. per Pack | 1 |
Product Type | Volumetric flask |
Class | A |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15821208
|
BRAND™
957482 |
10 mL |
518.00 DKK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
15841208
|
BRAND™
957484 |
25 mL |
518.00 DKK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
15851208
|
BRAND™
957485 |
50 mL |
528.00 DKK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
15861208
|
BRAND™
957487 |
100 mL |
528.00 DKK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
15871208
|
BRAND™
957488 |
200 mL |
585.00 DKK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
15891208
|
BRAND™
957490 |
500 mL |
692.00 DKK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
Description
- Manufactured from Boro 3.3 glass
- Acc. to DIN EN ISO 1042
- Delivered with individual USP conformity certificate
- Class A, DE-M marking
Specifications
Polypropylene | |
1 | |
A |
Borosilicate Glass (3.3) | |
Volumetric flask | |
Stopper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers, with Certificate