Learn More
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers, with Certificate
Amber glass to protect light-sensitive contents. Delivered with individual USP conformity certificate.
Brand: BRAND™ 957490
640.90 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from Boro 3.3 glass
- Acc. to DIN EN ISO 1042
- Delivered with individual USP conformity certificate
- Class A, DE-M marking
Specifications
Polypropylene | |
500 mL | |
1 | |
NS 19/26 | |
A |
19 mm | |
Borosilicate Glass (3.3) | |
Volumetric flask | |
±0.20 mL | |
Stopper |
Product Content Correction
Product Title
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers, with Certificate > Capacity: 500mL
Spot an opportunity for improvement?Share a Content Correction