Få mere at vide
AM 404-d4, TRC
Artikelnummer. 17937439
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17937439 1 mg
17929149 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17937439 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5758121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1185244-71-6
Molekylær formel C26H33D4NO2
Molekylvægt (g/mol) 399.6
Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxyphenyl-d4)-5,8,11,14-eicosatetraenamide, (all-Z)-N-(4-Hydroxyphenyl-d4)-5,8,11,14-eicosatetraenamide;
IUPAC navn (5Z,8Z,11Z,14Z)-N-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
SMIL OC1=C([2H])C([2H])=C(NC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)C([2H])=C1[2H]
Tekniske data
Kemisk navn eller materiale AM 404-d4
Mængde 1 mg
InChI formel InChI=1S/C26H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-26(29)27-24-20-22-25(28)23-21-24/h6-7,9-10,12-13,15-16,20-23,28H,2-5,8,11,14,17-19H2,1H3,(H,27,29)/b7-6-,10-9-,13-12-,16-15-/i20D,21D,22D,23D