missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AM-2099, MedChemExpress
Description
AM-2099 is a potent and selective inhibitor of voltage-gated sodium channel Nav1.7 with an IC50 of 0.16 μM for human Nav1.7.
Specifications
Specifications
| Kemisk navn eller materiale | AM-2099 |
| CAS | 1443373-17-8 |
| Farve | Off-White |
| Renhedsgrad noter | Research |
| Molekylær formel | C19H13F3N4O3S2 |
| Mængde | 50 mg |
| Opløselighedsinformation | DMSO : ≥ 150 mg/mL (321.57 mM) |
| SMIL | COC1=CC(C(F)(F)F)=CC=C1C2=NC=NC3=CC(S(=O)(NC4=NC=CS4)=O)=CC=C32 |
| Molekylvægt (g/mol) | 466.46 |
| Formel vægt | 466.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?