Learn More
VACUUBRAND™ Aluminum Reducing Pieces With Small KF Flanges
Brand:  VACUUBRAND™ 20669041
342.20 DKK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10099600
  • 364.00 DKK / Each
Estimated Shipment: 04-06-2024
to see stock.
Specifications
25/16 mm
Aluminum
40 mm
Solenoid-operated valves VV and VV C
1 Each
Reducing Pieces