Få mere at vide
Alprenolol, MedChemExpress
Artikelnummer. 16454607
Encompass_Preferred
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16454607 50 mg
16408786 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16454607 Leverandør Medchem Express Leverandørnr. HYB1517
MedChemExpress Alprenolol is a non-selective beta blocker as well as 5-HT1A receptor antagonist.
Alprenolol is a non-selective beta blocker as well as 5-HT1A receptor antagonist.
Tekniske data
Kemisk navn eller materiale Alprenolol
CAS 13655-52-2
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C15H23NO2
Mængde 50 mg
Synonym (RS)-Alprenolol dl-Alprenolol
Opløselighedsinformation DMSO : 100 mg/mL (401.04 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL OC(COC1=CC=CC=C1CC=C)CNC(C)C
Molekylvægt (g/mol) 249.35
Formel vægt 249.35
Procent renhed 98.04%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death