Få mere at vide
Alpha-Citral, TRC
Artikelnummer. 30007459
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007459 500 mg
17923025 100 mg
17990365 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007459 Leverandør Toronto Research Chemicals Leverandørnr. TRCC141275500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 141-27-5
Molekylær formel C10H16O
Molekylvægt (g/mol) 152.24
Synonym (2E)-3,7-Dimethyl-2,6-octadienal, (2E)-3,7-Dimethyl-2,6-octadien-1-al, (E)-3,7-Dimethyl-2,6-octadienal, (E)-3,7-Dimethyl-2,6-octadienal, (E)-Citral, (E)-Geranial, (E)-Neral, Citral a, Geranaldehyde, Geranial, trans-3,7-Dimethyl-2,6-octadien-1-al, trans-3,7-Dimethyl-2,6-octadienal, trans-Citral, trans-Geranial, beta-Geranial, beta-Neral;
IUPAC navn (2E)-3,7-dimethylocta-2,6-dienal
SMIL CC(=CCC\C(=C\C=O)\C)C
Tekniske data
Kemisk navn eller materiale alpha-Citral
Mængde 500 mg
InChI formel InChI=1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3/b10-7+