missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Alanine ethyl ester hydrochloride, 98%

CAS: 4244-84-2 | C5H12ClNO2 | 153.61 g/mol

459.00 DKK - 1715.00 DKK

Chemical Identifiers

CAS 4244-84-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012909
InChI Key RJCGNNHKSNIUAT-UHFFFAOYSA-N
Synonym ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl
PubChem CID 458475
IUPAC Name ethyl 3-aminopropanoate hydrochloride
SMILES Cl.CCOC(=O)CCN
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10676861
SDS View Documents
Thermo Scientific Chemicals
303060100
10 g Glass Bottle
459.00 DKK
10g
Please to purchase this item. Need a web account? Register with us today!
10432855
SDS View Documents
Thermo Scientific Chemicals
303060500
50 g Glass Bottle
1715.00 DKK
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals beta-Alanine ethyl ester hydrochloride, 98%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.