Få mere at vide
AHR 11652, TRC
Artikelnummer. 30133229
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133229 100 mg
30007667 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133229 Leverandør Toronto Research Chemicals Leverandørnr. TRCA435000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 241825-88-7
Molekylær formel C15H8BrNO3
Molekylvægt (g/mol) 330.13
Synonym 7-(4-Bromobenzoyl)-1H-indole-2,3-dione;
IUPAC navn 7-(4-bromobenzoyl)-1H-indole-2,3-dione
SMIL C1=CC(=C2C(=C1)C(=O)C(=O)N2)C(=O)C3=CC=C(C=C3)Br
Tekniske data
Kemisk navn eller materiale AHR 11652
Mængde 100 mg
InChI formel InChI=1S/C15H8BrNO3/c16-9-6-4-8(5-7-9)13(18)10-2-1-3-11-12(10)17-15(20)14(11)19/h1-7H,(H,17,19,20)
Formel vægt 328.9688