missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
AGI-5198, MedChemExpress
Beskrivelse
AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | AGI-5198 |
| CAS | 1355326-35-0 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C27H31FN4O2 |
| Mængde | 100 mg |
| Synonym | IDH-C35 |
| Opløselighedsinformation | DMF : ≥ 50 mg/mL (108.09 mM) ∣DMSO : 20.83 mg/mL (45.03 mM; Need ultrasonic) |
| SMIL | FC1=CC=CC(N(C(C2=CC=CC=C2C)C(NC3CCCCC3)=O)C(CN4C=CN=C4C)=O)=C1 |
| Molekylvægt (g/mol) | 462.56 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?