missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
AgBR1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67743-1-IG-150UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The AgBR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of mosquito AgBR1. This antibody recognizes mosquito antigen. The AgBR1 antibody has been validated for the following applications: WB, ELISA analysis.Tekniske data
| AgBR1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6034353 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, ELISA | |
| 2D1F10 | |
| Unconjugated | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Mosquito | |
| 40 kDa | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion