Få mere at vide
Adrafinil, TRC
Artikelnummer. 30009217
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009217 25 mg
17930515 5 mg
17953665 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009217 Leverandør Toronto Research Chemicals Leverandørnr. TRCA30150025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63547-13-7
Molekylær formel C15H15NO3S
Molekylvægt (g/mol) 289.35
Synonym Acetamide, 2-[(diphenylmethyl)sulfinyl]-N-hydroxy- (9CI, ACI), 2-[(Diphenylmethyl)sulfinyl]-N-hydroxyacetamide (ACI), 2-Benzhydrylsulfinyl-N-hydroxyacetamide, Adrafinil, CRL 40028, Olmifon
IUPAC navn 2-benzhydrylsulfinyl-N-hydroxyacetamide
SMIL ONC(=O)CS(=O)C(c1ccccc1)c2ccccc2
Tekniske data
Kemisk navn eller materiale Adrafinil
Mængde 25 mg
InChI formel InChI=1S/C15H15NO3S/c17-14(16-18)11-20(19)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15,18H,11H2,(H,16,17)
Formel vægt 289.0773