missing translation for 'onlineSavingsMsg'
Få mere at vide

Adoprazine, MedChemExpress

Artikelnummer. 16456168
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16456168 10 mg
16438767 5 mg
16432307 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16456168 Leverandør Medchem Express Leverandørnr. HY14782

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Adoprazine (SLV313) is a full 5-HT1A receptor agonist with a pEC50 of 9 at cloned h5-HT1A receptors. Adoprazine (SLV313) is a full D2 and D3 receptor antagonist with pA2s of 9.3 and 8.9 at hD2 and hD3 receptors, respectively. Adoprazine (SLV313) has the characteristics of atypical antipsychotics.

Adoprazine (SLV313) is a full 5-HT1A receptor agonist with a pEC50 of 9 at cloned h5-HT1A receptors. Adoprazine (SLV313) is a full D2 and D3 receptor antagonist with pA2s of 9.3 and 8.9 at hD2 and hD3 receptors, respectively. Adoprazine (SLV313) has the characteristics of atypical antipsychotics.

Tekniske data

Kemisk navn eller materiale Adoprazine
CAS 222551-17-9
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C24H24FN3O2
Mængde 10 mg
Synonym SLV313
Opløselighedsinformation DMSO : 50 mg/mL (123.32 mM; Need ultrasonic)
SMIL FC1=CC=C(C2=CN=CC(CN3CCN(C4=C5OCCOC5=CC=C4)CC3)=C2)C=C1
Molekylvægt (g/mol) 405.46
Formel vægt 405.46
Procent renhed 98.1%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.