Få mere at vide
Adomeglivant, MedChemExpress
Artikelnummer. 16490418
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16490418 5 mg
16483706 25 mg
16473887 50 mg
16457958 10 mg
16477746 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16490418 Leverandør Medchem Express Leverandørnr. HY19904
MedChemExpress Adomeglivant (LY2409021) is a potent, selective glucagon receptor (GluR) allosteric antagonist. Adomeglivant is widely used in the research for type 2 diabetes mellitus.
Adomeglivant (LY2409021) is a potent, selective glucagon receptor (GluR) allosteric antagonist. Adomeglivant is widely used in the research for type 2 diabetes mellitus.
Tekniske data
Kemisk navn eller materiale Adomeglivant
CAS 1488363-78-5
Farve White
Renhedsgrad noter Research
Molekylær formel C32H36F3NO4
Mængde 5 mg
Synonym LY2409021
Opløselighedsinformation DMSO : ≥ 100 mg/mL (179.98 mM)
SMIL O=C(O)CCNC(C1=CC=C([C@@H](OC2=CC(C)=C(C3=CC=C(C(C)(C)C)C=C3)C(C)=C2)CCC(F)(F)F)C=C1)=O
Molekylvægt (g/mol) 555.63
Formel vægt 555.63
Procent renhed 98.18%
Grad Research
Fysisk form Solid
Til brug med (applikation) Metabolism-sugar/lipid metabolism