Få mere at vide
AdipoRon, MedChemExpress
Artikelnummer. 16476868
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16476868 10 mg
16442937 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16476868 Leverandør Medchem Express Leverandørnr. HY15848
MedChemExpress AdipoRon is an orally active adiponectin receptor (AdipoR) agonist, binding to AdipoR1 and AdipoR2 with Kds of 1.8 and 3.1 μM, respectively.
AdipoRon is an orally active adiponectin receptor (AdipoR) agonist, binding to AdipoR1 and AdipoR2 with Kds of 1.8 and 3.1 μM, respectively.
Tekniske data
Kemisk navn eller materiale AdipoRon
CAS 924416-43-3
Farve White
Renhedsgrad noter Research
Molekylær formel C27H28N2O3
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 44 mg/mL (102.68 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(NC1CCN(CC2=CC=CC=C2)CC1)COC3=CC=C(C(C4=CC=CC=C4)=O)C=C3
Molekylvægt (g/mol) 428.52
Formel vægt 428.52
Procent renhed 96.13%
Grad Research
Fysisk form Powder
Til brug med (applikation) Metabolism-protein/nucleotide metabolism