Få mere at vide
Acridine-d9, TRC
Artikelnummer. 30133991
Skift visning
Mængde:
2 mg
4 mg
20 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133991 4 mg
30133259 2 mg
17929399 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30133991 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1909634MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34749-75-2
Molekylær formel C13 D9 N
Molekylvægt (g/mol) 188.27
Synonym Acridine D9, Perdeuterated acridine, Acridine-d9
IUPAC navn 1,2,3,4,5,6,7,8,9-nonadeuterioacridine
SMIL [2H]c1c([2H])c([2H])c2c([2H])c3c([2H])c([2H])c([2H])c([2H])c3nc2c1[2H]
Tekniske data
Kemisk navn eller materiale Acridine (D9)
Mængde 4 mg
InChI formel InChI=1S/C13H9N/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h1-9H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
Formel vægt 188.13