missing translation for 'onlineSavingsMsg'
Få mere at vide

Acid Yellow 17 (Technical Grade), TRC

Artikelnummer. 30135855
Change view
Click to view available options
Mængde:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30135855 250 g
30006118 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135855 Supplier Toronto Research Chemicals Supplier No. TRCA795235250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6359-98-4
Molekylær formel C16 H10 Cl2 N4 O7 S2 . 2 Na
Molekylvægt (g/mol) 551.289
Synonym C.I. Acid Yellow 17, C.I. Acid Yellow 17, Disodium Salt, 1437 Yellow, Acid Brilliant Yellow 2G, Acid Fast Yellow 2G, Acid Fast Yellow 2GL, Acid Leather Yellow 2GL, Acid Light Yellow 2G, Acid Yellow 2G, Acidic Light Yellow 2G, Acidine Fast Yellow 2GD, Amacid Fast Light Yellow G, Amacid Light Yellow 2G, Ambicid Yellow 2G, Apocid Light Yellow 2GX, Apollo Nylon Fast Light Yellow 2G, Belacid Fast Light Yellow 2G, Best Acid Light Yellow 2G, Bucacid Light Yellow 2G, C.I. 13075, C.I. 18965, C.I. Food Yellow 5, Calcocid Fast Yellow 2G, Cetil Light Yellow 2G, Colocid Light Fast Yellow 2G, Conacid Yellow AL, Concorde Acid Yellow GC, Covanyl Light Yellow 2G, Covasol Yellow W 1007, Daiwa Acid Yellow 2G, Diacid Light Yellow 2G, Dinacid Fast Yellow 2G, Dinacid Light Fast Yellow 2G, Dyacid Yellow 2G, Dycosacid Brilliant Yellow 2G, Dynacid Yellow 2G, Egacid Yellow 3G, Erio Fast Flavine 3G, Everacid Light Yellow GC, Fabracid Yellow S 2GL, Fast Light Yellow 2G, Fastlite Yellow 2G, Fenalan Yellow G, Food Yellow No. 5, Hexacol Yellow 2G, Hidacid Light Yellow 2G, Ichoacid Yellow G, Indacid Yellow 2G, Intracid Fast Yellow 2GL, Java Light Yellow 2GN, KCA Fast Light Yellow 5GL, Kayacyl Yellow GC, Kayacyl Yellow GG, Kayaku Acid Fast Yellow GG, Kemacid Yellow 2G, Kiton Fast Yellow 2GL, Light Acid Yellow 2G, Lighthouse Fast Yellow 2G, Lissamine Fast Yellow 2G, Metamine Fast Light Yellow 2GX, Mitsui Acid Fast Yellow G, Mitsui Acid Fast Yellow G Conc., Multacid Yellow 2G, NSC 324981, Nankai Fast Light Yellow 2G, Naphthazine Light Yellow 2G, Neran Brilliant Yellow 2G, New Lemon EMBL, Nylomine Acid Yellow P 4G, Pontacyl Light Yellow GX, Pyrazolone Yellow 2J, Ravi Acid Yellow 2G, Rifa Acid Fast Yellow 2G, Romexal Yellow 2G, Sandolan Yellow E 2GL, Simacid Yellow, Triacid Light Yellow 2GL, Unitertracid Light Yellow 2G, Vibra Color Yellow AYE 17, Vicoacid Light Fast Yellow 2G, Vondacid Light Yellow 2GL, Water Yellow 350053, Xylene Light Yellow 2G, Xylene Light Yellow 3G, Xylene Light Yellow N 2G;
IUPAC navn disodium;2,5-dichloro-4-[5-hydroxy-3-methyl-4-[(E)-(4-sulfonatophenyl)diazenyl]pyrazol-1-yl]benzenesulfonate
SMIL [Na+].[Na+].Cc1nn(c(O)c1N=Nc2ccc(cc2)S(=O)(=O)[O-])c3cc(Cl)c(cc3Cl)S(=O)(=O)[O-]

Specifications

Kemisk navn eller materiale Acid Yellow 17
InChI formel InChI=1S/C16H12Cl2N4O7S2.2Na/c1-8-15(20-19-9-2-4-10(5-3-9)30(24,25)26)16(23)22(21-8)13-6-12(18)14(7-11(13)17)31(27,28)29;;/h2-7,23H,1H3,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2
Mængde 250 g
Formel vægt 549.916
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.