missing translation for 'onlineSavingsMsg'
Få mere at vide

Acid Blue 9 (Technical Grade), TRC

Artikelnummer. 30134965
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134965 50 mg
30132647 100 mg
30012596 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30134965 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18981550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2650-18-2
Molekylær formel C37H42N4O9S3
Molekylvægt (g/mol) 782.95
Synonym Alphazurine FG (6CI), Benzenemethanaminium, N-ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](2-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfo-, hydroxide, inner salt, diammonium salt, Benzenemethanaminium, N-ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](2-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfo-, inner salt, diammonium salt (9CI), C.I. Acid Blue 9, diammonium salt (8CI), 11388 Blue, A.F. Blue No. 1, AE 85, Acid Blue 9-39003, Acid Blue AE-SF, Acid Blue FG, Acid Brilliant Blue FCF, Acid Brilliant Blue FG, Acid Enoglaucine, Acid Sky Blue A, Acilan Turquoise Blue AE, Aizen Brilliant Blue FCF, Alphazurine, Alphazurine FGND, Amacid Blue FG Conc, Basacid Blue NB 754, Basacid Blue NB 757, Basacid Green 21L, Best Acid Brilliant Blue FCF, Blue 17608, Blue 205, Blue No. 205, Brilliant Blue FCF 307047, Brilliant Blue FCF 36021, Brilliant Blue FCF 37005, Brilliant Blue FCF 37012, Brilliant Blue FCF 37013, Brilliant Blue FCF 90130, Brilliant Blue FDF 37002, Bucacid Azure Blue, C.I. 2650182, C.I. Acid Blue 9, C.I. Acid Blue 9 ammonium salt, Calcocid Blue 2G, Calcocid Blue EG, Cogilor Blue 512.12, Colocid Brilliant Blue C, D and C Blue No. 4, D&C Blue No. 4, Daiwa Blue 1, Daiwa Blue 109H, Daiwa Blue 1WB, Dinacid Patent Blue AE, Disulphine Lake Blue EG, Duasyn Acid Blue AE, Duasyn Acid Blue AE 85, Duasyn Acid Blue AE-SF, Dyacid Turquoise AN, Dycosacid Turquoise Blue GZ, Edicol Supra Blue E 6, Erioglaucine, Erioglaucine A, Erioglaucine E, Eriosky Blue, Eurocert Brilliant Blue FCF 312507, FD & C Blue No 1 308001, FD&C Blue No. 1 37002, FD&C Blue No. 1 37005, FD&C Blue No. 1 37012, FD&C Blue No. 1 37013, FD&C Blue No. 1 90130, Fenazo Blue XR, H.K. Formula No. K 7117, Hexacol Brilliant Blue, Hidacid Azure Blue, Iragon Blue ABL 9, Japan Blue 205, Kemacid Brilliant Blue FCF, Keyacid Blue FG, Kiton Blue AR, Kiton Pure Blue L, Lerui Acid Blue FG, Maple Brilliant Blue FCF, NSC 4318, NSC 5018, Naphthazine Brilliant Blue AE, Necol Brilliant Blue FCF, Neelicol Brilliant Blue FCF, Neolan Blue E-A, Neptune Blue BRA, Orient Water Blue 9, Pacid Blue, Patent Blue 2Y, Patent Blue AE, Peacock Blue X 1756, Pro-Jet Blue OAM, Rakuto Brilliant Blue FCF, Ravi Patent Blue AE, Sandolan Blue E-VSG, Simacid Blue A, Triacid Blue AE, Usacert FD & C Blue No 1 310501, Vicoacid Blue 9, Vitasyn Blue AE 85, Water Blue 310502, Water Blue 9, Xylene Blue VSG
IUPAC navn diazanium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
SMIL CC/[N+](CC1=CC(S(=O)([O-])=O)=CC=C1)=C2C=C/C(C=C/2)=C(C3=CC=CC=C3S(=O)([O-])=O)/C4=CC=C(N(CC5=CC(S(=O)([O-])=O)=CC=C5)CC)C=C4.[NH4+].[NH4+]

Tekniske data

Kemisk navn eller materiale Acid Blue 9
Mængde 50 mg
InChI formel InChI=1S/C37H36N2O9S3.2H3N/c1-3-38(25-27-9-7-11-33(23-27)49(40,41)42)31-19-15-29(16-20-31)37(35-13-5-6-14-36(35)51(46,47)48)30-17-21-32(22-18-30)39(4-2)26-28-10-8-12-34(24-28)50(43,44)45;;/h5-24H,3-4,25-26H2,1-2H3,(H2-,40,41,42,43,44,45,46,47,48);2*1H3
Formel vægt 782.2114
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.