Få mere at vide
Aceclofenac, 99%, Thermo Scientific™
667.92 DKK
Kemiske identifikatorer
| CAS | 89796-99-6 |
|---|---|
| Molekylær formel | C16H13Cl2NO4 |
| Molekylvægt (g/mol) | 354.183 |
| MDL nummer | MFCD00864296 |
| InChI nøgle | MNIPYSSQXLZQLJ-UHFFFAOYSA-N |
| Synonym | aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish |
| PubChem CID | 71771 |
| ChEBI | CHEBI:31159 |
| IUPAC navn | 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| SMIL | C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl |
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
A non-steroidal, anti-inflammatory drug (NSAID) with potent inhibitory activity in several models of inflammation.Kemiske identifikatorer
| 89796-99-6 | |
| 354.183 | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 71771 | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid |
| C16H13Cl2NO4 | |
| MFCD00864296 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| CHEBI:31159 | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl |
Tekniske data
| Aceclofenac | |
| C16H13Cl2NO4 | |
| UN2811 | |
| aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish | |
| MNIPYSSQXLZQLJ-UHFFFAOYSA-N | |
| 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid | |
| 71771 | |
| 354.18 |
| 89796-99-6 | |
| MFCD00864296 | |
| 14,22 | |
| Soluble in DMSO | |
| C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl | |
| 354.183 | |
| CHEBI:31159 | |
| 99% |
Sikkerhed og håndtering
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P310-P305+P351+P338-P330-P501c
H301-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : CY1577900
missing translation for 'tsca' : No
Anbefalet opbevaring : Ambient temperatures
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.