Få mere at vide
Abemaciclib methanesulfonate, MedChemExpress
Beskrivelse
Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Abemaciclib methanesulfonate |
| CAS | 1231930-82-7 |
| Farve | Light Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C28H36F2N8O3S |
| Mængde | 100 mg |
| Synonym | LY2835219 methanesulfonate |
| Opløselighedsinformation | H2O : 125 mg/mL (207.40 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (16.59 mM; ultrasonic and warming and heat to 80°C) |
| SMIL | CC(N1C2=CC(C3=NC(NC4=NC=C(CN5CCN(CC)CC5)C=C4)=NC=C3F)=CC(F)=C2N=C1C)C.CS(=O)(O)=O |
| Molekylvægt (g/mol) | 602.7 |
| Vis mere |