Få mere at vide
Tygon A-60-G Chemical Dispensing Tubing
Specielt formuleret slange giver fremragende modstandsdygtighed over for uorganiske væsker
Brand: Tygon 50106-62
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. Outstanding resistance to heat, UV, and ozone. Use in temperatures ranging from -60°C to 135°C (-75°F to 275°F). High flexural fatigue strength—ideal for use in peristaltic pumps. Heat sealable—no fittings needed to join tubingTekniske data
7/16 in. | |
9/16 in. | |
Black |
Tygon A-60-G | |
50 ft. |
Korrektion af produktindhold
Produkttitel
Tygon A-60-G Chemical Dispensing Tubing > 9/16 in.,7/16 in.
Ser du en mulighed for forbedring?Del en indholdskorrektion