Få mere at vide
9-Dehydrotestosterone, TRC
Artikelnummer. 30011998
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011998 100 mg
30008801 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011998 Leverandør Toronto Research Chemicals Leverandørnr. TRCD230205100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2398-99-4
Molekylær formel C19H26O2
Molekylvægt (g/mol) 286.41
Synonym 17beta-Hydroxyandrosta-4,9(11)-dien-3-one, 4,9(11)-Androstadiene-17beta-ol-3-one, 9(11)-Dehydrotestosterone, NSC 30932, U 3435
SMIL C[C@]12CC=C3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O
Tekniske data
Kemisk navn eller materiale 9-Dehydrotestosterone
Mængde 100 mg
InChI formel InChI=1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h8,11,14-15,17,21H,3-7,9-10H2,1-2H3/t14-,15-,17-,18-,19-/m0/s1
Formel vægt 286.1933