Learn More
Hamilton™ 800 Microliter Syringes, No Needle Included
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 250 µL.
Brand:  Hamilton™ 7646-01
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11550052
  • 1255.00 DKK / Each
Estimated Shipment: 17-05-2024
to see stock.
Description
Description
  • Ideal for dispensing volumes from 0.5 µL up to 10 µL
  • Syringe holder is permanently attached to the syringe barrel
Specifications
Specifications
250 μL