Få mere at vide
8-anti-Ipratropium Bromide, TRC
Artikelnummer. 30133237
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133237 25 mg
30009963 2.5 mg
30006193 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133237 Leverandør Toronto Research Chemicals Leverandørnr. TRCI74050525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58073-59-9
Molekylær formel C20 H30 N O3 . Br
Molekylvægt (g/mol) 412.36
Synonym 8-Azoniabicyclo[3.2.1]octane, 3-(3-hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-, bromide (1:1), (3-endo,8-anti)-, 8-Azoniabicyclo[3.2.1]octane, 3-(3-hydroxy-1-oxo-2-phenylpropoxy)-8-methyl-8-(1-methylethyl)-, bromide, (3-endo,8-anti)- (9CI);
SMIL OCC(C1=CC=CC=C1)C(O[C@H]2CC3CCC(C2)[N@@+]3(C(C)C)C)=O.[Br-]
Tekniske data
Kemisk navn eller materiale (1R,3r,5S,8s)-3-(((2RS)-3-hydroxy-2-phenylpropanoyl)oxy)-8-methyl-8-(1-methylethyl)-8-azoniabicyclo(3.2.1)octane
Mængde 25 mg
InChI formel InChI=1S/C20H30NO3.BrH/c1-14(2)21(3)16-9-10-17(21)12-18(11-16)24-20(23)19(13-22)15-7-5-4-6-8-15;/h4-8,14,16-19,22H,9-13H2,1-3H3;1H/q+1;/p-1
Formel vægt 411.1409