Få mere at vide
7-(4-Bromobenzoyl)-1,3-dihydro-2H-indol-2-one, TRC
Artikelnummer. 30003861
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003861 50 mg
30009458 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003861 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68019050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 91713-91-6
Molekylær formel C15 H10 Br N O2
Molekylvægt (g/mol) 316.15
Synonym 7-(4-Bromobenzoyl)-1,3-dihydro-2H-indol-2-one, AHR 10240
IUPAC navn 7-(4-bromobenzoyl)-1,3-dihydroindol-2-one
SMIL Brc1ccc(cc1)C(=O)c2cccc3CC(=O)Nc23
Tekniske data
Kemisk navn eller materiale 7-(4-Bromobenzoyl)-1,3-dihydro-2H-indol-2-one
Mængde 50 mg
InChI formel InChI=1S/C15H10BrNO2/c16-11-6-4-9(5-7-11)15(19)12-3-1-2-10-8-13(18)17-14(10)12/h1-7H,8H2,(H,17,18)
Formel vægt 314.9895