Få mere at vide
6Beta-Hydroxy Progesterone (Contains up to 20% 17Alpha-Hydroxy Progesterone), TRC
Artikelnummer. 30006555
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006555 5 mg
30004415 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006555 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9522455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 604-19-3
Molekylær formel C21 H30 O3
Molekylvægt (g/mol) 330.46
Synonym Pregn-4-ene-3,20-dione, 6-hydroxy-, (6β)-, Pregn-4-ene-3,20-dione, 6β-hydroxy- (8CI), Progesterone, 6β-hydroxy- (6CI), (6β)-6-Hydroxypregn-4-ene-3,20-dione, 4-Pregnen-6β-ol-3,20-dione, 6β-Hydroxypregn-4-en-3,20-dione, 6β-Hydroxypregn-4-ene-3,20-dione, 6β-Hydroxyprogesterone, U 27605
IUPAC navn (6R,8S,9S,10R,13S,14S,17S)-17-acetyl-6-hydroxy-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMIL CC(=O)[C@H]1CC[C@H]2[C@@H]3C[C@@H](O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale Progesterone 6-beta-Hydroxy
Mængde 5 mg
InChI formel InChI=1S/C21H30O3/c1-12(22)15-4-5-16-14-11-19(24)18-10-13(23)6-8-21(18,3)17(14)7-9-20(15,16)2/h10,14-17,19,24H,4-9,11H2,1-3H3/t14-,15+,16-,17-,19+,20+,21+/m0/s1
Formel vægt 330.2195