Få mere at vide
6-Methoxy-2-naphthaleneacetic Acid-d3 (Desmethyl Naproxen-d3), TRC
Artikelnummer. 30466163
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466163 1 mg
30471371 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466163 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2646971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189427-82-4
Molekylær formel C13H9D3O3
Molekylvægt (g/mol) 219.25
Synonym 2-(6-Methoxy-d3-2-naphthyl)acetic Acid, 6-Methoxy-d3-2-naphthylacetic Acid, 6-MNA-d3, BRL 10720-d3, Desmethyl Naproxen-d3
SMIL O=C(O)CC1=CC2=C(C=C1)C=C(OC([2H])([2H])[2H])C=C2
Tekniske data
Kemisk navn eller materiale Desmethyl Naproxen-d3
Mængde 1 mg
InChI formel InChI=1S/C13H12O3/c1-16-12-5-4-10-6-9(7-13(14)15)2-3-11(10)8-12/h2-6,8H,7H2,1H3,(H,14,15)/i1D3