Få mere at vide
6-Fluoro DL-DOPA Hydrobromide Salt, TRC
Artikelnummer. 30009361
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009361 10 mg
30006008 1 mg
30003986 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009361 Leverandør Toronto Research Chemicals Leverandørnr. TRCF59155010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1323114-61-9
Molekylær formel C9 H10 F N O4 . Br H
Molekylvægt (g/mol) 296.09
Synonym Tyrosine, 2-fluoro-5-hydroxy-, hydrobromide (1:1), 6-Fluoro DL-DOPA hydrobromide salt, 2-Fluoro-5-hydroxy-DL-tyrosine hydrobromide
IUPAC navn 2-amino-3-(2-fluoro-4,5-dihydroxyphenyl)propanoic acid;hydrobromide
SMIL Br.NC(Cc1cc(O)c(O)cc1F)C(=O)O
Tekniske data
Kemisk navn eller materiale 2-Fluoro-5-hydroxy-DL-tyrosine Hydrobromide
InChI formel InChI=1S/C9H10FNO4.BrH/c10-5-3-8(13)7(12)2-4(5)1-6(11)9(14)15;/h2-3,6,12-13H,1,11H2,(H,14,15);1H
Mængde 10 mg
Formel vægt 294.9855