Få mere at vide
6-Desacetyl Palbociclib, TRC
Artikelnummer. 30133348
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133348 5 mg
30133141 50 mg
30012703 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133348 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2887185MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 571190-22-2
Molekylær formel C22H27N7O
Molekylvægt (g/mol) 405.5
Synonym 8-Cyclopentyl-5-methyl-2-[[(5-piperazin-1-ylpyridin-2-yl)]amino]-8H-pyrido[2,3-d]pyrimidin-7-one;
SMIL CC1=CC(=O)N(C2CCCC2)c3nc(Nc4ccc(cn4)N5CCNCC5)ncc13
Tekniske data
Kemisk navn eller materiale 6-Desacetyl Palbociclib
Analyt- eller komponentnavne 6-Desacetyl Palbociclib
Forsendelsestilstand Room Temperature
Løsningstype Neat
Industry Type Pharmaceutical