missing translation for 'onlineSavingsMsg'
Få mere at vide

6,7,8,9,10,11-Hexahydrocyclooct[b]indole, 97%, Thermo Scientific™

Artikelnummer. 11493028
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
11493028 5 g 5g
11483028 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Product Code. 11493028 Supplier Thermo Scientific Alfa Aesar Supplier No. A18758.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22793-63-1
Molekylær formel C14H17N
Molekylvægt (g/mol) 199.30
MDL nummer MFCD00086346
InChI nøgle TYLZSGIRJRUUSQ-UHFFFAOYSA-N
Synonym 6,7,8,9,10,11-hexahydro-5h-cycloocta b indole, 5h,6h,7h,8h,9h,10h,11h-cycloocta b indole, 5h-cyclooct b indole,6,7,8,9,10,11-hexahydro, acmc-20am0o, maybridge1_006281, tylzsgirjruusq-uhfffaoysa, 6,7,8,9,10,11-hexahydro-5h-cyclooct b indole, 6,7,8,9,10,11-hexahydrocycloocta 1,2-b indole, 6,7,8,9,10,11-hexahydrocycloocta 2,1-b indole
PubChem CID 272686
IUPAC navn 5H,6H,7H,8H,9H,10H,11H-cycloocta[b]indole
SMIL C1CCCC2=C(CC1)NC1=CC=CC=C21

Specifications

Kemisk navn eller materiale 6,7,8,9,10,11-Hexahydrocyclooct[b]indole
Smeltepunkt 72°C to 76°C
Mængde 5 g
Formel vægt 199.3
Procent renhed 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.