Få mere at vide
5H-Pyrido[3,2-b]azepine-6,9-(7H,8H)-dione, TRC
Artikelnummer. 30425413
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425413 250 mg
30422992 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30425413 Leverandør Toronto Research Chemicals Leverandørnr. TRCP991710250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 676596-63-7
Molekylær formel C9 H8 N2 O2
Molekylvægt (g/mol) 176.17
Synonym 5H-Pyrido[3,2-b]azepine-6,9-dione, 7,8-dihydro-, 5H-Pyrido[3,2-b]azepine-6,9(7H,8H)-dione, 7,8-Dihydro-5H-pyrido[3,2-b]azepine-6,9-dione
IUPAC navn 7,8-dihydro-5H-pyrido[3,2-b]azepine-6,9-dione
SMIL O=C1CCC(=O)c2ncccc2N1
Tekniske data
Kemisk navn eller materiale 5H-Pyrido[3,2-b]azepine-6,9-(7H,8H)-dione
Mængde 250 mg
InChI formel InChI=1S/C9H8N2O2/c12-7-3-4-8(13)11-6-2-1-5-10-9(6)7/h1-2,5H,3-4H2,(H,11,13)
Formel vægt 176.0586