Få mere at vide
5alpha-Androstenone, TRC
Artikelnummer. 30007806
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007806 100 mg
30012861 5 mg
30010828 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007806 Leverandør Toronto Research Chemicals Leverandørnr. TRCA637775100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18339-16-7
Molekylær formel C19 H28 O
Molekylvægt (g/mol) 272.42
Synonym Androst-16-en-3-one, (5α)- (9CI, ACI), (5α)-Androst-16-en-3-one (ACI), 5α-Androst-16-en-3-one (7CI, 8CI), Androstenone (6CI), 16,(5α)-Androsten-3-one, 5α-Androst-16-ene-3-one, 5α-Androstenone, E 282
IUPAC navn (5S,8R,9S,10S,13R,14S)-10,13-dimethyl-1,2,4,5,6,7,8,9,11,12,14,15-dodecahydrocyclopenta[a]phenanthren-3-one
SMIL C[C@]12CC[C@H]3[C@@H](CC[C@H]4CC(=O)CC[C@]34C)[C@@H]1CC=C2
Tekniske data
Kemisk navn eller materiale 5alpha-Androstenone
Mængde 100 mg
InChI formel InChI=1S/C19H28O/c1-18-9-3-4-16(18)15-6-5-13-12-14(20)7-11-19(13,2)17(15)8-10-18/h3,9,13,15-17H,4-8,10-12H2,1-2H3/t13-,15-,16-,17-,18-,19-/m0/s1
Formel vægt 272.214