Få mere at vide
(5alpha)-2'H-Androst-2-eno[3,2-c]pyrazol-17-one, TRC
Artikelnummer. 30138733
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138733 500 mg
30003870 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138733 Leverandør Toronto Research Chemicals Leverandørnr. TRCA637515500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 129545-93-3
Molekylær formel C20H28N2O
Molekylvægt (g/mol) 312.45
Synonym 2'H-Androst-2-eno[3,2-c]pyrazol-17-one, (5α)-, (5α)-2'H-Androst-2-eno[3,2-c]pyrazol-17-one, 17-Ketoprostanozol
IUPAC navn (1S,2S,10S,13R,14S,18S)-2,18-dimethyl-6,7-diazapentacyclo[11.7.0.02,10.04,8.014,18]icosa-4(8),5-dien-17-one
SMIL C[C@]12Cc3cn[nH]c3C[C@@H]1CC[C@@H]4[C@@H]2CC[C@@]5(C)[C@H]4CCC5=O
Tekniske data
Kemisk navn eller materiale (5a)-2'H-Androst-2-eno[3,2-c]pyrazol-17-one
Mængde 500 mg
InChI formel InChI=1S/C20H28N2O/c1-19-8-7-16-14(15(19)5-6-18(19)23)4-3-13-9-17-12(11-21-22-17)10-20(13,16)2/h11,13-16H,3-10H2,1-2H3,(H,21,22)/t13-,14-,15-,16-,19-,20-/m0/s1
Formel vægt 312.2202