Learn More
Cellpath System II Hex™ Tissue Processing Cassettes (Hoppered)
Pre-loaded into hoppers for use with automated and carousel type printers. CellPath System II Hex Tissue Processing Cassettes (Hoppered) feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
1434.69 DKK
Specifications
Quantity | 1500/Pk. |
---|---|
Material | Acetyl Resin |
Product Type | Tissue Processing Cassette in Hopper tube |
Description
- Pre-loaded into hoppers for use with automated and carousel type cassette printers
- Manufactured from high–grade virgin acetyl resin
- Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
- Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
- Extra thick side walls increases rigidity and stability for high quality sectioning
- Designed to suit Thermo Printmate and Micro-writer cassette–writing machines
- Available in 15 stock colors which color code different types of workload or applications
Specifications
1500/Pk. | |
Acetyl Resin | |
Tissue Processing Cassette in Hopper tube |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.