Få mere at vide
5-(Trifluoromethyl)indolin-2-one, TRC
Artikelnummer. 30453960
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453960 50 mg
30450498 100 mg
30451145 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30453960 Leverandør Toronto Research Chemicals Leverandørnr. TRCT89964350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 71293-62-4
Molekylær formel C9H6NOF3
Molekylvægt (g/mol) 201.15
Synonym 1,3-Dihydro-5-(trifluoromethyl)-2H-indol-2-one (ACI), 5-(Trifluoromethyl)-1,3-dihydroindol-2-one, 5-(Trifluoromethyl)-2-oxindole, 5-(Trifluoromethyl)indolin-2-one, 5-(Trifluoromethyl)oxindole
IUPAC navn 5-(trifluoromethyl)indolin-2-one
SMIL O=C1NC2=C(C=C(C(F)(F)F)C=C2)C1
Tekniske data
Kemisk navn eller materiale 5-(Trifluoromethyl)indolin-2-one
Mængde 50 mg
InChI formel InChI=1S/C9H6F3NO/c10-9(11,12)6-1-2-7-5(3-6)4-8(14)13-7/h1-3H,4H2,(H,13,14)
Formel vægt 201.04