Få mere at vide
5-Oxomefruside, TRC
Artikelnummer. 30003936
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003936 1 mg
30005819 10 mg
30004617 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003936 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8578501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14599-36-1
Molekylær formel C13H17ClN2O6S2
Molekylvægt (g/mol) 396.87
Synonym 4-Chloro-N1-methyl-N1-[(tetrahydro-2-methyl-5-oxo-2-furanyl)methyl]-1,3-benzenedisulfonamide, 4-Chloro-N1-methyl-N1-(tetrahydro-2-methyl-5-oxofurfuryl)-m-benzenedisulfonamide, 2-[N-(4'-Chloro-3'-sulfamoylbenzenesulfono)-N-methyl-2-aminomethyl]-2-methyl-5-oxotetrahydrofuran, 4-Chloro-N1-methyl-N1-(2-methyl-5-oxotetrahydrofurfuryl)-m-benzenedisulfonamide, Mefruside Lactone, N-(4-Chloro-3-sulfamoylbenzenesulfonyl)-N-methyl-2-aminomethyl-2-methyl-5-oxotetrahydrofuran
IUPAC navn 4-chloro-1-N-methyl-1-N-[(2-methyl-5-oxooxolan-2-yl)methyl]benzene-1,3-disulfonamide
SMIL O=[S](N(C)CC(CCC1=O)(O1)C)(C2=CC([S](N)(=O)=O)=C(Cl)C=C2)=O
Tekniske data
Kemisk navn eller materiale 5-Oxomefruside
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C13H17ClN2O6S2/c1-13(6-5-12(17)22-13)8-16(2)24(20,21)9-3-4-10(14)11(7-9)23(15,18)19/h3-4,7H,5-6,8H2,1-2H3,(H2,15,18,19)
Formel vægt 396.0217
Procent renhed >95%