Få mere at vide
5-Oxo-1,3-dioxolane-4,4-diacetic Acid, TRC
Artikelnummer. 30412968
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412968 1 g
30423296 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30412968 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8555001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 144-16-1
Molekylær formel C7H8O7
Molekylvægt (g/mol) 204.13
Synonym beta-(Hydroxymethoxy)-tricarballylic acid gamma-Lactone, Anhydromethylenecitric Acid, Methylenecitric Acid, NSC 68614
IUPAC navn 2-[4-(carboxymethyl)-5-oxo-1,3-dioxolan-4-yl]acetic acid
SMIL OC(=O)CC1(CC(=O)O)OCOC1=O
Tekniske data
Kemisk navn eller materiale 5-Oxo-1,3-dioxolane-4,4-diacetic Acid
Mængde 1 g
InChI formel InChI=1S/C7H8O7/c8-4(9)1-7(2-5(10)11)6(12)13-3-14-7/h1-3H2,(H,8,9)(H,10,11)
Formel vægt 204.027