Få mere at vide
5-Methylbenz[a]anthracene, TRC
Artikelnummer. 30133155
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133155 100 mg
30132713 50 mg
30007614 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133155 Leverandør Toronto Research Chemicals Leverandørnr. TRCM288795100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2319-96-2
Molekylær formel C19H14
Molekylvægt (g/mol) 242.31
Synonym 5-Monomethylbenz[a]anthracene, 5-Methylbenzo[a]anthracene;
SMIL Cc1cc2cc3ccccc3cc2c4ccccc14
Tekniske data
Kemisk navn eller materiale 5-Methylbenz[a]anthracene
Mængde 100 mg
InChI formel InChI=1S/C19H14/c1-13-10-16-11-14-6-2-3-7-15(14)12-19(16)18-9-5-4-8-17(13)18/h2-12H,1H3
Formel vægt 242.1096