Få mere at vide
5-Methyl-2'-O-methylcytidine, TRC
Artikelnummer. 30008534
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008534 250 mg
30007878 100 mg
30006366 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008534 Leverandør Toronto Research Chemicals Leverandørnr. TRCM263410250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 113886-70-7
Molekylær formel C11 H17 N3 O5
Molekylvægt (g/mol) 271.27
Synonym 4-Amino-1-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one
IUPAC navn 4-amino-1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxy-tetrahydrofuran-2-yl]-5-methyl-pyrimidin-2-one
SMIL CO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C)C(=NC2=O)N
Tekniske data
Kemisk navn eller materiale 5-Methyl-2'-O-methylcytidine
Mængde 250 mg
InChI formel InChI=1S/C11H17N3O5/c1-5-3-14(11(17)13-9(5)12)10-8(18-2)7(16)6(4-15)19-10/h3,6-8,10,15-16H,4H2,1-2H3,(H2,12,13,17)/t6-,7-,8-,10-/m1/s1
Formel vægt 271.1168