Få mere at vide
5-(Hydroxymethyl)-4-methyl-2-(4-trifluoromethylphenyl)thiazole, TRC
Artikelnummer. 30132487
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132487 500 mg
30004331 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132487 Leverandør Toronto Research Chemicals Leverandørnr. TRCH947795500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 317318-96-0
Molekylær formel C12 H10 F3 N O S
Molekylvægt (g/mol) 273.2741
Synonym 4-Methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolemethanol, [4-Methyl-2-(4-trifluoromethylphenyl)thiazol-5-yl]methanol, [4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl]methanol
IUPAC navn [4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl]methanol
SMIL Cc1nc(sc1CO)c2ccc(cc2)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 5-(Hydroxymethyl)-4-methyl-2-(4-trifluoromethylphenyl)thiazole
Mængde 500 mg
InChI formel InChI=1S/C12H10F3NOS/c1-7-10(6-17)18-11(16-7)8-2-4-9(5-3-8)12(13,14)15/h2-5,17H,6H2,1H3
Formel vægt 273.0435