Få mere at vide
5-(Hydroxymethyl)-2’-deoxycytidine, TRC
Artikelnummer. 30011269
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011269 10 mg
30007885 5 mg
30005125 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011269 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94663010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7226-77-9
Molekylær formel C10 H15 N3 O5
Molekylvægt (g/mol) 257.24
Synonym 2'-deoxy-5-(hydroxymethyl)cytidine, 5-(Hydroxymethyl)deoxycytidine;
IUPAC navn 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(hydroxymethyl)pyrimidin-2-one
SMIL NC1=NC(=O)N(C=C1CO)[C@H]2C[C@H](O)[C@@H](CO)O2
Tekniske data
Kemisk navn eller materiale 5-(Hydroxymethyl)-2'-deoxycytidine
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C10H15N3O5/c11-9-5(3-14)2-13(10(17)12-9)8-1-6(16)7(4-15)18-8/h2,6-8,14-16H,1,3-4H2,(H2,11,12,17)/t6-,7+,8+/m0/s1
Formel vægt 257.1012
Procent renhed >95%