Få mere at vide
5-Chloro-3-(methylamino)sulfamoyl-2-carboxylic Acid Methyl Ester, TRC
Artikelnummer. 30364789
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364789 10 mg
30363104 50 mg
30368091 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364789 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36904810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70374-37-7
Molekylær formel C7H8ClNO4S2
Molekylvægt (g/mol) 269.73
IUPAC navn methyl 5-chloro-3-(methylsulfamoyl)thiophene-2-carboxylate
SMIL CNS(=O)(=O)c1cc(Cl)sc1C(=O)OC
Tekniske data
Kemisk navn eller materiale 5-Chloro-3-(methylamino)sulfamoyl-2-carboxylic Acid Methyl Ester
Mængde 10 mg
InChI formel InChI=1S/C7H8ClNO4S2/c1-9-15(11,12)4-3-5(8)14-6(4)7(10)13-2/h3,9H,1-2H3
Formel vægt 268.9583