Få mere at vide
5-Chloro-2-hydroxybenzophenone, TRC
Artikelnummer. 17961895
Skift visning
Mængde:
100 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17961895 5 g
17902595 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17961895 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3682505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85-19-8
Molekylær formel C13 H9 Cl O2
Molekylvægt (g/mol) 232.66
Synonym 5-Chloro-2-hydroxy-benzophenone, (2-Hydroxy-5-chlorophenyl)phenylmethanone, (5-Chloro-2-hydroxyphenyl)phenylmethanone, 2-Benzoyl-4-chlorophenol, 2-Hydroxy-5-chlorobenzophenone, 3-Chloro-6-hydroxybenzophenone, 5-Chloro-2-hydroxybenzophenone, Benzophenone 7, Ch 11, DOW HCB, NSC 33407, UV Absorber NL/5
IUPAC navn (5-chloro-2-hydroxy-phenyl)-phenyl-methanone
SMIL Oc1ccc(Cl)cc1C(=O)c2ccccc2
Tekniske data
Kemisk navn eller materiale 5-Chloro-2-hydroxybenzophenone
Mængde 5 g
InChI formel InChI=1S/C13H9ClO2/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8,15H
Formel vægt 232.0291